Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH4
Entrez Gene ID
508912
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Gene Aliases
CNIH-4
Chromosome Location
Chr.16: 27529034 - 27543276 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034338.2 NP_001029510.1 3 - 4 331 105
GenBank mRNA BC102152.1 - 3 - 4 333 105

View Full Product Details