Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLF2
Entrez Gene ID
513843
Gene Name
myeloid leukemia factor 2
Gene Aliases
-
Chromosome Location
Chr.5: 103691259 - 103696646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034456.1 NP_001029628.1 1 - 2 147 120
GenBank mRNA BC104523.1 - 1 - 2 147 120

View Full Product Details