Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP2A
Entrez Gene ID
523847
Gene Name
charged multivesicular body protein 2A
Gene Aliases
-
Chromosome Location
Chr.18: 65775981 - 65778680 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034550.2 NP_001029722.1 1 - 2 127 85
GenBank mRNA BC102258.1 - 1 - 2 128 85

View Full Product Details