Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHMP2A
Entrez Gene ID
523847
Gene Name
charged multivesicular body protein 2A
Gene Aliases
-
Chromosome Location
Chr.18: 65775981 - 65778680 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034550.2 NP_001029722.1 4 - 5 650 96
GenBank mRNA BC102258.1 - 4 - 5 651 96

View Full Product Details