Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD46
Entrez Gene ID
534898
Gene Name
ankyrin repeat domain 46
Gene Aliases
-
Chromosome Location
Chr.14: 63800447 - 63868341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034619.1 NP_001029791.1 2 - 3 186 123
GenBank mRNA BC104580.1 - 2 - 3 186 123

View Full Product Details