Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP1
Entrez Gene ID
538615
Gene Name
THAP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.27: 37544991 - 37553519 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034648.1 NP_001029820.1 1 - 2 301 88
GenBank mRNA BC102409.1 - 1 - 2 301 88

View Full Product Details