Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED9
Entrez Gene ID
618580
Gene Name
transmembrane emp24 protein transport domain containing 9
Gene Aliases
-
Chromosome Location
Chr.7: 39117554 - 39121523 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034812.2 NP_001029984.1 4 - 5 574 86
GenBank mRNA BC102145.1 - 4 - 5 575 86
BT021063.1 - 4 - 5 566 86

View Full Product Details