Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD1
Entrez Gene ID
510774
Gene Name
abhydrolase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 72508608 - 72515678 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035094.2 NP_001030266.1 7 - 8 877 65
GenBank mRNA BC102483.1 - 7 - 8 879 65

View Full Product Details