Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM1
Entrez Gene ID
527379
Gene Name
zinc finger MYM-type containing 1
Gene Aliases
ZNF262
Chromosome Location
Chr.3: 110595281 - 110623082 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205509.3 NP_001192438.2 1 - 2 182 151
GenBank mRNA BC102673.1 - 1 - 2 180 150

View Full Product Details