Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAMDC
Entrez Gene ID
533224
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, C29H11orf67
Chromosome Location
Chr.29: 18154903 - 18189531 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037473.2 NP_001032550.1 3 - 4 309 80
GenBank mRNA BC108191.1 - 3 - 4 311 80

View Full Product Details