Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf76
Entrez Gene ID
613457
Gene Name
chromosome 2 C2orf76 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 71107880 - 71214464 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037618.2 NP_001032707.1 5 - 6 397 114
GenBank mRNA BC109869.1 - 5 - 6 398 114

View Full Product Details