Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCP3
Entrez Gene ID
615896
Gene Name
synaptonemal complex protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 65541079 - 65548288 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040588.2 NP_001035678.1 6 - 7 492 102
GenBank mRNA BC102433.1 - 6 - 7 493 102

View Full Product Details