Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL11
Entrez Gene ID
512745
Gene Name
ribosomal protein L11
Gene Aliases
-
Chromosome Location
Chr.2: 129194243 - 129198106 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075581.2 NP_001069049.1 1 - 1 414 99
GenBank mRNA BC102524.1 - 1 - 1 416 99

View Full Product Details