Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP10
Entrez Gene ID
613957
Gene Name
GTP binding protein 10
Gene Aliases
-
Chromosome Location
Chr.4: 74533001 - 74559892 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080314.2 NP_001073783.1 5 - 6 615 154
GenBank mRNA BC105245.1 - 5 - 6 617 154

View Full Product Details