Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2H
Entrez Gene ID
539313
Gene Name
ubiquitin conjugating enzyme E2 H
Gene Aliases
E2-20K, UBCH2
Chromosome Location
Chr.4: 93646643 - 93748860 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037614.2 NP_001032703.1 3 - 4 648 111
GenBank mRNA BC109502.1 - 3 - 4 649 111

View Full Product Details