Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D20
Entrez Gene ID
504543
Gene Name
TBC1 domain family member 20
Gene Aliases
-
Chromosome Location
Chr.13: 60581690 - 60600670 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038029.1 NP_001033118.1 7 - 8 1062 89
GenBank mRNA BC111319.1 - 7 - 8 1062 89

View Full Product Details