Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN4
Entrez Gene ID
505789
Gene Name
centrin 4
Gene Aliases
-
Chromosome Location
Chr.17: 34949127 - 34953268 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038039.1 NP_001033128.1 1 - 2 116 135
GenBank mRNA BC109577.1 - 1 - 2 116 135

View Full Product Details