Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAP29
Entrez Gene ID
511018
Gene Name
B cell receptor associated protein 29
Gene Aliases
-
Chromosome Location
Chr.4: 48591446 - 48644225 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038075.1 NP_001033164.1 1 - 2 59 79
GenBank mRNA BC110030.1 - 1 - 2 59 79

View Full Product Details