Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL2
Entrez Gene ID
511349
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
-
Chromosome Location
Chr.29: 43227243 - 43235043 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038079.2 NP_001033168.1 1 - 2 364 138
GenBank mRNA BC111133.1 - 1 - 2 365 138

View Full Product Details