Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYC1
Entrez Gene ID
512500
Gene Name
cytochrome c1
Gene Aliases
-
Chromosome Location
Chr.14: 738111 - 740532 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038090.2 NP_001033179.1 4 - 5 650 68
GenBank mRNA BC109917.1 - 4 - 5 651 68
BC148921.1 - 3 - 4 1199 68

View Full Product Details