Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP45
Entrez Gene ID
617350
Gene Name
cilia and flagella associated protein 45
Gene Aliases
CCDC19
Chromosome Location
Chr.3: 9783427 - 9812756 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038219.2 NP_001033308.2 5 - 6 671 63
GenBank mRNA BC109498.1 - 5 - 6 672 63

View Full Product Details