Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED1
Entrez Gene ID
538144
Gene Name
transmembrane p24 trafficking protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 15298595 - 15301886 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038563.2 NP_001033652.1 3 - 4 484 65
GenBank mRNA BC110006.1 - 3 - 4 485 65

View Full Product Details