Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB4A
Entrez Gene ID
615849
Gene Name
RAB4A, member RAS oncogene family
Gene Aliases
RAB4
Chromosome Location
Chr.28: 1197453 - 1268190 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038685.2 NP_001033774.1 4 - 5 333 77
GenBank mRNA BC110198.1 - 4 - 5 334 77

View Full Product Details