Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZWINT
Entrez Gene ID
514564
Gene Name
ZW10 interacting kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.26: 2809079 - 2813939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040529.2 NP_001035619.1 2 - 3 165 66
GenBank mRNA BC110184.1 - 2 - 3 166 66

View Full Product Details