Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPZ1
Entrez Gene ID
519496
Gene Name
spermatogenic leucine zipper 1
Gene Aliases
-
Chromosome Location
Chr.20: 276052 - 283737 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040540.2 NP_001035630.1 1 - 1 1446 140
GenBank mRNA BC109808.1 - 1 - 1 1447 140

View Full Product Details