Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED27
Entrez Gene ID
525389
Gene Name
mediator complex subunit 27
Gene Aliases
CRSP8
Chromosome Location
Chr.11: 101923662 - 102165161 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040546.2 NP_001035636.1 7 - 8 822 81
GenBank mRNA BC109883.1 - 7 - 8 823 81

View Full Product Details