Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBX3
Entrez Gene ID
539222
Gene Name
PBX homeobox 3
Gene Aliases
-
Chromosome Location
Chr.11: 96551862 - 96777501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046453.1 NP_001039918.1 6 - 7 1110 80
GenBank mRNA BC110144.1 - 6 - 7 1110 80

View Full Product Details