Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
BNIP3
Entrez Gene ID
615342
Gene Name
BCL2 interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.26: 51221338 - 51225301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076366.1 NP_001069834.1 - 445 64
GenBank mRNA BC110012.1 - - 445 64

View Full Product Details