Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGB3BP
Entrez Gene ID
614469
Gene Name
integrin subunit beta 3 binding protein
Gene Aliases
-
Chromosome Location
Chr.3: 82052235 - 82157908 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079619.2 NP_001073087.1 4 - 5 327 154
GenBank mRNA BC112592.1 - 4 - 5 328 154

View Full Product Details