Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6H4orf17
Entrez Gene ID
616552
Gene Name
chromosome 6 C4orf17 homolog
Gene Aliases
-
Chromosome Location
Chr.6: 25111320 - 25137614 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080323.2 NP_001073792.1 3 - 4 669 137
GenBank mRNA BC111327.1 - 3 - 4 670 137

View Full Product Details