Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCOLCE
Entrez Gene ID
504471
Gene Name
procollagen C-endopeptidase enhancer
Gene Aliases
-
Chromosome Location
Chr.25: 35954408 - 35959426 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045888.1 NP_001039353.1 4 - 5 685 65
GenBank mRNA BC113211.1 - 4 - 5 685 65

View Full Product Details