Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
508494
Gene Name
solute carrier family 22 (organic anion transporter), member 9
Gene Aliases
SLC22A25
Chromosome Location
Chr.29: 41663877 - 41689883 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046006.1 NP_001039471.1 9 - 10 1634 67
GenBank mRNA BC112499.1 - 9 - 10 1634 67

View Full Product Details