Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSRC1
Entrez Gene ID
541250
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
-
Chromosome Location
Chr.3: 34125570 - 34129207 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046501.1 NP_001039966.1 5 - 6 766 108
GenBank mRNA BC114140.1 - 5 - 6 766 108

View Full Product Details