Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB2A
Entrez Gene ID
508373
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
RAB2
Chromosome Location
Chr.14: 26180786 - 26253380 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075354.1 NP_001068822.1 2 - 3 130 99
GenBank mRNA BC118271.1 - 2 - 3 130 99

View Full Product Details