Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BID
Entrez Gene ID
510373
Gene Name
BH3 interacting domain death agonist
Gene Aliases
-
Chromosome Location
Chr.5: 109135720 - 109154339 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075446.2 NP_001068914.1 1 - 2 164 118
GenBank mRNA BC118359.1 - 1 - 2 166 118

View Full Product Details