Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNG
Entrez Gene ID
511381
Gene Name
uracil DNA glycosylase
Gene Aliases
-
Chromosome Location
Chr.17: 63961374 - 63976150 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075513.2 NP_001068981.1 1 - 2 437 104
GenBank mRNA BC118447.1 - 1 - 2 438 104

View Full Product Details