Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCD
Entrez Gene ID
513272
Gene Name
IQ motif containing D
Gene Aliases
-
Chromosome Location
Chr.17: 61202727 - 61225586 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075606.1 NP_001069074.1 3 - 4 1139 70
GenBank mRNA BC118347.1 - 3 - 4 1139 70

View Full Product Details