Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFP
Entrez Gene ID
539605
Gene Name
complement factor properdin
Gene Aliases
-
Chromosome Location
Chr.X: 85997325 - 86002710 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076178.2 NP_001069646.1 3 - 4 476 101
GenBank mRNA BC118432.1 - 3 - 4 478 101

View Full Product Details