Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRMS1
Entrez Gene ID
615952
Gene Name
BRMS1 transcriptional repressor and anoikis regulator
Gene Aliases
-
Chromosome Location
Chr.29: 44389726 - 44396831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076404.1 NP_001069872.1 9 - 10 998 85
GenBank mRNA BC116061.1 - 9 - 10 998 85

View Full Product Details