Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JKAMP
Entrez Gene ID
509181
Gene Name
JNK1/MAPK8 associated membrane protein
Gene Aliases
JAMP
Chromosome Location
Chr.10: 71759901 - 71782137 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080247.1 NP_001073716.1 2 - 3 99 105
GenBank mRNA BC116146.1 - 2 - 3 99 105

View Full Product Details