Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POMGNT2
Entrez Gene ID
497030
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Gene Aliases
AGO61, C22H3orf39, C3orf39, GTDC2
Chromosome Location
Chr.22: 15060105 - 15080384 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011682.2 NP_001011682.1 1 - 2 183 99
GenBank mRNA BC123822.1 - 1 - 2 183 99

View Full Product Details