Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLG2
Entrez Gene ID
505152
Gene Name
DNA polymerase gamma 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.19: 48657930 - 48677642 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075191.1 NP_001068659.1 6 - 7 1261 86
GenBank mRNA BC120380.1 - 6 - 7 1261 86

View Full Product Details