Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLG2
Entrez Gene ID
505152
Gene Name
DNA polymerase gamma 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.19: 48657930 - 48677642 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075191.1 NP_001068659.1 7 - 8 1364 134
GenBank mRNA BC120380.1 - 7 - 8 1364 134

View Full Product Details