Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIKE1
Entrez Gene ID
506848
Gene Name
suppressor of IKBKE 1
Gene Aliases
SIKE
Chromosome Location
Chr.3: 28556617 - 28562489 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075281.2 NP_001068749.1 1 - 2 252 147
GenBank mRNA BC120197.1 - 1 - 2 253 147

View Full Product Details