Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC32
Entrez Gene ID
506935
Gene Name
coiled-coil domain containing 32
Gene Aliases
C10H15orf57
Chromosome Location
Chr.10: 36151423 - 36161279 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075286.1 NP_001068754.1 2 - 3 475 99
GenBank mRNA BC120381.1 - 2 - 3 475 99

View Full Product Details