Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YRDC
Entrez Gene ID
512062
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 108108908 - 108112891 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075550.1 NP_001069018.1 2 - 3 599 64
GenBank mRNA BC120307.1 - 2 - 3 599 64

View Full Product Details