Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf4
Entrez Gene ID
515243
Gene Name
chromosome 5 C12orf4 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 105596049 - 105636160 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075704.1 NP_001069172.1 12 - 13 1600 102
GenBank mRNA BC119869.1 - 12 - 13 1600 102

View Full Product Details