Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIB
Entrez Gene ID
525408
Gene Name
NFKB inhibitor beta
Gene Aliases
-
Chromosome Location
Chr.18: 48614347 - 48624970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075872.1 NP_001069340.1 3 - 4 685 92
GenBank mRNA BC122726.1 - 3 - 4 685 92

View Full Product Details