Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSUN3
Entrez Gene ID
533791
Gene Name
NOP2/Sun RNA methyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.1: 38360629 - 38420857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076001.2 NP_001069469.1 2 - 3 135 141
GenBank mRNA BC120181.1 - 2 - 3 136 141

View Full Product Details