Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2U1
Entrez Gene ID
535227
Gene Name
cytochrome P450, family 2, subfamily U, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.6: 17261762 - 17282966 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076050.2 NP_001069518.1 2 - 3 1185 74
GenBank mRNA BC122663.1 - 2 - 3 1186 74

View Full Product Details